Learn More
Hamilton™ Polilyte Lab Temp
Specifications
Specifications
| Connector Types | DIN |
| Type | Polilyte Lab Temp |
| Includes | Temperature sensor |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | -10°C to +100°C |
| Length (Metric) | 120 mm |