Learn More
Invitrogen™ PolarScreen™ ER Beta Competitor Assay Kit, Green
Product Code. 15353826
missing translation for 'orderingAttributeHoverText'
Quantity:
2500 Assays
800 Assays
missing translation for 'unitSize'
800 units
Pack of 2500
Product Code. 15353826
missing translation for 'mfr': Invitrogen™ A15889
The PolarScreen™ ER Beta Competitor Assay Kit provides a sensitive and efficient method for high-throughput, fluorescence polarization-based screening of potential estrogen receptor (ER) beta ligands.
The PolarScreen™ ER Beta Competitor Assay Kit provides a sensitive and efficient method for high-throughput, fluorescence polarization-based screening of potential estrogen receptor (ER) beta ligands. The kit uses insect cell-expressed, full-length, native (untagged) human ER beta (Cat. No. A15664) and a novel, high-affinity, fluorescent estrogen ligand (Fluormone™ ES2) in a homogenous, mix-and-read assay format.
This kit produces a green color read-out; a 'red' kit is also available (A15891). Both kits suitably determine the IC50s of competitive compounds. Color choice depends on the spectral properties of the compounds to be screened and the available instrumentation. For additional information or assistance email drugdiscoverytech@lifetech.com.
The PolarScreen™ ER Beta Competitor Assay Kit is:
  • Complete & ready-to-use out of the box—all reagents included, just add your test compounds
  • High throughput compatible—20 μL fluorescence polarization assays in a 384-well plate
  • Convenient—available in bulk sizes for larger screening needs
How the Assay Works
Full-length ER beta is added to a fluorescent estrogen ligand to form an ER-Fluormone™ ES2 complex. This complex is added to individual test compounds in multiwell plates. To determine relative affinity, the assay measures fluorescence polarization. Competing test compounds will displace the Fluormone™ ES2 ligand from ER beta, permitting it to tumble rapidly and resulting in low polarization values. A shift in the fluorescence polarization value in the presence of test compound is used to determine the relative affinity of test compounds for ER beta.
All components are manufactured under the strictest quality parameters. This product's performance, per the Certificat of Analysis, is guaranteed for 6 months after purchase. Contact us for bulk quantities or for assistance with other nuclear receptor assay needs, from assay development to library screening and beyond.
TRUSTED_SUSTAINABILITY
Specifications
Color Green
Description PolarScreen™ ER Beta Competitor Assay Kit, 2500 Assays, Green, 384-Well Plate Format, Fluorescence Polarization Technique, Estrogen Receptor Beta Nuclear Receptor Target Gene, For Pharma & Biopharma, Target & Lead Identification
Includes 1 Tube of ER beta human recombinant protein, full length, 1 tube of Fluromone™ ES2, 1 tube of ES2 screening buffer
Quantity 2500 Assays
Regulatory Status For research use only (RUO)
Packaging 384-well Plate
Content And Storage • Estrogen Receptor (ER) Beta Recombinant Protein, Full Length (store at -68°C to -85°C)
• Fluromone™ ES2 (store at -5°C to -30° C)
• ES2 Screening Buffer (store at room temperature)
Assay Category Biochemical Assay
Druggable Target Nuclear Receptors
Readout Fluorescence Polarization
Product Line PolarScreen
For Use With (Application) Nuclear Receptor Assay, Fluorescence Polarization
Ligand ER beta
Product Type Assay Kit
Target Molecule Estrogen Receptor Beta Nuclear Receptor