Learn More
Trajan Scientific and Medical™ Replacement Plunger for Fixed Needle Thermo RSH Autosampler Syringes
SGE syringe replacement plungers for gas tight syringes
Brand:  Trajan Scientific and Medical™ 032831
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 13189185
  • 376.00 NOK / Each
Estimated Shipment: 22-07-2024
Description
  • Replacement plungers for gas tight syringes
  • Designed to match specific syringe barrel to maintain gas tight seal
Specifications
Specifications
PTFE (Plunger)
Gastight
250 μL