Learn More
Trajan Scientific and Medical™ Replacement Plunger for Fixed Needle Thermo RSH Autosampler Syringes
SGE syringe replacement plungers for gas tight syringes
362.00 NOK - 407.00 NOK
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
13149185
|
Trajan Scientific and Medical™
032810 |
10 μL |
392.00 NOK
Each |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13159185
|
Trajan Scientific and Medical™
032815 |
25 μL |
362.00 NOK
Each |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13169185
|
Trajan Scientific and Medical™
032821 |
50 μL |
362.00 NOK
Each |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13179185
|
Trajan Scientific and Medical™
032825 |
100 μL |
376.00 NOK
Each |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13189185
|
Trajan Scientific and Medical™
032831 |
250 μL |
376.00 NOK
Each |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13199185
|
Trajan Scientific and Medical™
032835 |
500 μL |
407.00 NOK
Each |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Replacement plungers for gas tight syringes
- Designed to match specific syringe barrel to maintain gas tight seal
Specifications
PTFE (Plunger) | |
Gastight |