Learn More
Memmert™ Plug-In Tube Extension for SN75plus, Straight
Product Code. 17508775 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
SN75plus Sterilizer
Unit Size:
Each
Product Code. 17508775
Brand: Memmert™ SN75plusB29718
Only null left
Add to basket
Add to basket
Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.
  • Accessory
  • Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
  • Straight
Specifications
Diameter (Metric) Outside 60.3 mm
For Use With (Equipment) SN75plus Sterilizer