Learn More
Memmert™ Plug-In Tube Extension for SN750plus, Straight
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
SN750plus Sterilizer
Unit Size:
Each
Description
- Accessory
- Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
- Straight
Specifications
Specifications
| Diameter (Metric) Outside | 60.3 mm |
| For Use With (Equipment) | SN750plus Sterilizer |