Learn More
Memmert™ Plug-In Tube Extension for SF75, Straight
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
SF75 Sterilizer
Unit Size:
Each
Description
- Accessory
- Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
- Straight
Specifications
Specifications
| Diameter (Metric) Outside | 60.3 mm |
| For Use With (Equipment) | SF75 Sterilizer |