Learn More
SI Analytics™ Connection Cables and Plugs
Product Code. 10152071 Shop All SI Analytics Products
Change view
Click to view available options
Length (Metric) Cable:
1 m
10 m
2 m
5 m
Type:
K 1 A
K 10 A
K2X
K5A
L 1 A
L 1 BNC
L 1 N
L 1 NN
L 1 R
L 2 A
Plug Cable Combination
Unit Size:
Each
Product Code. Length (Metric) Cable Type unitSize
10152071 2 m K2X Each
10001072 1 m L 1 NN Each
10535553 5 m K 10 A Each
10326932 1 m L 1 N Each
10604671 1 m L 1 BNC Each
10406812 1 m K 1 A Each
10305111 1 m L 1 R Each
10684661 1 m L 1 A Each
10466165 2 m L 2 A Each
10652832 5 m Plug Cable Combination Each
10559371 10 m K5A Each
11 options
Product Code. 10152071
Brand: SI Analytics™ 285120028
Only null left
Add to basket
Add to basket
SI Analytics™ Connection Cables and Plugs are available in various combinations of electrode socket, instrument plug and cable length.
  • Coaxial plug for pH, redox, ammonia and sodium combination electrodes, pH and redox single electrodes as well as reference electrodes in Plus series. The L and LB series plugs are compatible. The LB sockets are matching colors with the BlueLine electrodes
  • Electrode plug for reference electrodes from the predecessor series
  • Plug for conductivity measuring cells with temperature sensor and oxygen cells, for older models
TRUSTED_SUSTAINABILITY
Specifications
Connector Types Coaxial Electrode Plug
Type K2X
Includes 2m Coaxial Cable, Free End
Length (Metric) Cable 2 m