missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLOD3 Rabbit anti-Human, Mouse, Rat, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1696.32 NOK - 5307.92 NOK
Specifications
| Antigen | PLOD3 |
|---|---|
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | CoraLite 594 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17731379
|
Proteintech
CL594-11027-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17639926
|
Proteintech
CL594-11027-100UL |
100 μL |
5307.92 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PLOD3 Polyclonal antibody apecifically detetects PLOD3 antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence.Specifications
| PLOD3 | |
| Polyclonal | |
| Liquid | |
| Human, Mouse, Rat | |
| BC011674 | |
| PLOD3 | |
| IgG | |
| Antigen affinity purification |
| Immunofluorescence | |
| CoraLite 594 | |
| Rabbit | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| 8985 | |
| PLOD3 fusion protein Ag1480 | |
| Primary | |
| Store at -20°C. Avoid exposure to light. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title