missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLOD3 Rabbit anti-Human, Mouse, Rat, CoraLite 594, Polyclonal, Proteintech
Shop All Proteintech ProductsDescription
PLOD3 Polyclonal antibody apecifically detetects PLOD3 antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence.
Specifications
Specifications
| Antigen | PLOD3 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 1000 μg/mL |
| Conjugate | CoraLite 594 |
| Formulation | PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. |
| Gene Accession No. | BC011674 |
| Gene Symbols | PLOD3 |
| Host Species | Rabbit |
| Immunogen | PLOD3 fusion protein Ag1480 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?