missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLDN Mouse anti-Human, Mouse, Rat, Clone: 4F4C11, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | PLDN |
|---|---|
| Clone | 4F4C11 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16848545
|
Proteintech
66558-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16838545
|
Proteintech
66558-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene may play a role in intracellular vesicle trafficking. It interacts with Syntaxin 13 which mediates intracellular membrane fusion. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.Specifications
| PLDN | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| PA, PALLID, Pallid protein homolog, Pallidin, pallidin homolog (mouse), PLDN | |
| Bloc1s6 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 4F4C11 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Mouse, Rat, Human | |
| Q4V8A6, Q9R0C0, Q9UL45 | |
| 18457, 26258, 317630 | |
| PLDN Fusion Protein Ag18095 | |
| Primary | |
| -20°C | |
| BLOC1S6 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title