missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLDN Mouse anti-Human, Mouse, Rat, Clone: 4F4C11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66558-1-IG-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene may play a role in intracellular vesicle trafficking. It interacts with Syntaxin 13 which mediates intracellular membrane fusion. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.Specifications
| PLDN | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q4V8A6, Q9R0C0, Q9UL45 | |
| Bloc1s6 | |
| PLDN Fusion Protein Ag18095 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 4F4C11 | |
| Unconjugated | |
| BLOC1S6 | |
| PA, PALLID, Pallid protein homolog, Pallidin, pallidin homolog (mouse), PLDN | |
| Mouse | |
| Protein G | |
| RUO | |
| 18457, 26258, 317630 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction