Learn More
Eppendorf™ Plate Carriers for Buckets
Product Code. 15748379 Shop All Eppendorf Products
Change view
Click to view available options
Type:
Plate Carrier for High-capacity Bucket
Plate Carrier for Plate/Tube Bucket
Capacity:
6/24 (MTP)\n2/8 (DWP)
7/28 (MTP)\n2/8 (DWP)
Unit Size:
Each
Product Code. Type Capacity unitSize
15748379 Plate Carrier for Plate/Tube Bucket 7/28 (MTP)\n2/8 (DWP) Each
15684614 Plate Carrier for High-capacity Bucket 6/24 (MTP)\n2/8 (DWP) Each
2 options
Product Code. 15748379
Brand: Eppendorf™ 5920705008
Carriers designed for MTP, DWP, PCR plates. Available for high-capacity buckets and plate/tube buckets
  • Number of tubes per adapter/per rotor: 7/28(MTP)\n2/8(DWP) or 6/24 (MTP)\n2/8 (DWP)
  • Set of 2 pcs.
TRUSTED_SUSTAINABILITY
Specifications
Type Plate Carrier for Plate/Tube Bucket
For Use With (Equipment) for MTP, DWP, PCR plates
Capacity 7/28 (MTP)\n2/8 (DWP)