Learn More
Fisherbrand™ Plastic PP Two-Part Syringes, Luer Lock
Product Code. p-7113855 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
2 mL
5 mL
10 mL
20 mL
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
12921031 10 mL Pack of 100
12941031 20 mL Pack of 100
12901031 5 mL Pack of 100
12981021 2 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12921031
Only null left
Add to basket
Add to basket
HPLC certified, non-sterile, two-part disposable syringes made of solvent robust polypropylene.
  • Plastic Disposable Syringe
  • Polypropylene
  • Luer Lock
  • Two-part, all-plastic construction eliminates the need for rubber or synthetic plunger gaskets
  • No silicone or oil lubricant is required in the barrel
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Volume (Metric) 10 mL
Connection Type Luer Lock