Learn More
Fisherbrand™ Plain Glass Plates with Bonded Spacers
Product Code. 11887643 Shop All Fisherbrand Products
Change view
Click to view available options
Includes:
0.5mm bonded spacers
0.75mm bonded spacers
1.5mm Bonded Spacers
2.0mm Bonded Spacers
Unit Size:
Pack of 2
Product Code. Includes unitSize
11887643 1.5mm Bonded Spacers Pack of 2
11857643 0.5mm bonded spacers Pack of 2
11867643 0.75mm bonded spacers Pack of 2
11897643 2.0mm Bonded Spacers Pack of 2
4 options
Product Code. 11887643
Only null left
Add to basket
Add to basket
Accessory for use with OmniPAGE mini vertical Electrophoresis systems.
  • Accessory for use with OmniPAGE mini vertical Electrophoresis systems
Specifications
Type Plain Glass Plate
Gel Dimensions 100 x 100 mm
Includes 1.5mm Bonded Spacers