Description
- With a redesigned piston assembly system and high quality piston grease, PIPETMAN™ G offers hours of comfortable pipetting
- PIPETMAN™ G relies on the legendary accuracy, precision, and reliability of PIPETMAN™ with improved pipetting comfort
- Made of PVDF and stainless steel
- Offers premium durability
Specifications
Specifications
| Certifications/Compliance | Declaration of Conformity |
| Autoclavable | Not Autoclavable |
| Color | Red |
| Material | Metal Ejector |
| Model | P12x10G |
| Number of Channels | 12 |
| Product Line | Pipetman G |
| Type | Manual Air Displacement Pipette |
| Volume (Metric) | 1 to 10 μL |
| Warranty | 3 Years |
| Show More |