missing translation for 'onlineSavingsMsg'
Learn More

Piperidine-1-thiocarboxamide, 98%, Thermo Scientific™

Product Code. 11313166
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313166 5 g 5g
11303166 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313166 Supplier Thermo Scientific Alfa Aesar Supplier No. L12445.06
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14294-09-8
Molecular Formula C6H12N2S
Molecular Weight (g/mol) 144.236
MDL Number MFCD00041230
InChI Key UERQMZVFUXRQOD-UHFFFAOYSA-N
Synonym 1-piperidinethiocarboxamide, 1-piperidinecarbothioamide, aminopiperidylmethane-1-thione, acmc-20aos3, piperidine thiocarboxamide, 1-thiocarbamyl piperidine, 1-piperidinethiocarboximide, 1-piperi-dinecarbothioamide, piperidine-1-thiocarboxamide, 1-piperidinecarbothioamide #
PubChem CID 1551232
IUPAC Name piperidine-1-carbothioamide
SMILES C1CCN(CC1)C(=S)N

Specifications

Melting Point 126°C to 130°C
Quantity 5 g
Beilstein 383640
Formula Weight 144.24
Percent Purity 98%
Chemical Name or Material Piperidine-1-thiocarboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.