accessibility menu, dialog, popup

UserName

CAS RN 14294-09-8

IUPAC Name: piperidine-1-carbothioamide
Synonyms: 1-piperidinethiocarboxamide
Molecular Weight (g/mol): 144.24
Molecular Formula: C6H12N2S
InChi Key: UERQMZVFUXRQOD-UHFFFAOYSA-N
SMILES: NC(=S)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results