Learn More
Thermo Scientific™ TSAT (tris-(succinimidyl)aminotriacetate)
Shop All Thermo Scientific Products
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Thermo Scientific™ Pierce TSAT is tris(succinimidyl)aminotriacetate, a novel trifunctional crosslinker that contains three short spacers arms linked to a central nitrogen atom, each ending in an amine-reactive NHS-ester group.
Highlights:
- Reactive group: maleimide
- Reactive toward: sulfhydryl groups
Specifications
Specifications
| Chemical Reactivity | Amine-Amine |
| Cleavable | No |
| Description | TSAT |
| Molecular Weight (g/mol) | 482.36 |
| PEGylated | No |
| Spacer Arm Length | 4.2 Å |
| Content And Storage | Upon receipt store at 4°C protected from moisture. |
| Cell Permeability | Yes |
| Shipping Condition | Ambient |
| Product Line | Pierce |
| Show More |