Learn More
Thermo Scientific™ Pierce™ Protease Inhibitor Tablets, EDTA-free
Shop All Thermo Scientific ProductsDescription
Pierce Protease Inhibitor Tablets protect proteins from proteolytic degradation through inhibition of the serine-proteases, cysteine-proteases, aspartic acid-proteases and aminopeptidases that are typically present in cellular lysate samples. The tablets contain AEBSF, aprotinin, bestatin, E-64, leupeptin and pepstatin A. Stable for storage at 4°C for up to 12 months, each mini or large tablet tablet is sufficient for 10mL or 50mL of solution, respectively, and is suitable for reagent preparation prior to protein extraction from tissue and cultured cells.
Highlights:
Specifications
Specifications
| Includes | 1 vial of inhibitor tablets |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | protein purification, western blotting |
| Sufficient For | 1 Tablet is Sufficient for 50 mL |
| Quantity | 2 Tablets |