missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospholipase C beta 2 Rabbit anti-Human, Primate, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1705.00 NOK - 4235.00 NOK
Specifications
| Antigen | Phospholipase C beta 2 |
|---|---|
| Dilution | Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence |
| Applications | Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18347822
|
Novus Biologicals
NBP3-11952-25UL |
25 μg |
1705.00 NOK
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18329793
|
Bio-Techne
NBP3-11952-100UL |
100 μg |
4235.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phospholipase C beta 2 Polyclonal antibody specifically detects Phospholipase C beta 2 in Human, Primate samples. It is validated for Immunohistochemistry, ImmunofluorescenceSpecifications
| Phospholipase C beta 2 | |
| Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism, Wnt Signaling Pathway | |
| 100 ul in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per ml BSA and 50% glycerol. | |
| 5330 | |
| Primary | |
| Store at -20C. |
| Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Primate | |
| EC 3.1.4.11, FLJ38135, Phosphoinositide phospholipase C-beta-2,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-2, phospholipase C, beta 2, Phospholipase C-beta-2, PLC-beta-2 | |
| Peptide corresponding to amino acid residues from the C-terminal region of Phospholipase C beta 2. (UniProt: Q00722) | |
| Antigen Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title