missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-STAT5A (Tyr694) Rabbit anti-Human, CoraLite 650, Clone: 2P21, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech CL650-80115-20UL
Additional Details : Weight : 0.00010kg
Description
Phospho-STAT5A (Tyr694) antibody apecifically detetects Phospho-STAT5A (Tyr694) antigen in Human Samples. It is validated for Flow Cytometry.Specifications
Phospho-STAT5A (Tyr694) | |
2P21 | |
CoraLite 650 | |
BC027036 | |
Rabbit | |
Protein A purification | |
Primary | |
Human | |
Liquid |
Flow Cytometry | |
1000 ug/mL | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
STAT5A | |
Peptide | |
20 μL | |
6776 | |
Store at -20°C. Avoid exposure to light. | |
IgG |