missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-STAT5A (Tyr694) Rabbit anti-Human, CoraLite 650, Clone: 2P21, Proteintech
Rabbit Recombinant Antibody
1415.13 NOK - 4925.53 NOK
Specifications
Antigen | Phospho-STAT5A (Tyr694) |
---|---|
Clone | 2P21 |
Applications | Flow Cytometry |
Conjugate | CoraLite 650 |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17701489
|
Proteintech
CL650-80115-20UL |
20 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
17640067
|
Proteintech
CL650-80115-100UL |
100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Phospho-STAT5A (Tyr694) antibody apecifically detetects Phospho-STAT5A (Tyr694) antigen in Human Samples. It is validated for Flow Cytometry.Specifications
Phospho-STAT5A (Tyr694) | |
Flow Cytometry | |
Liquid | |
Human | |
BC027036 | |
STAT5A | |
IgG | |
Protein A purification |
2P21 | |
CoraLite 650 | |
Rabbit | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
6776 | |
Peptide | |
Primary | |
Store at -20°C. Avoid exposure to light. |