missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-AKT1 (Ser473) Rabbit anti-Human, Mouse, CoraLite 594, Clone: 2M10, Proteintech
Rabbit Recombinant Antibody
1651.35 NOK - 5307.92 NOK
Specifications
| Antigen | Phospho-AKT1 (Ser473) |
|---|---|
| Clone | 2M10 |
| Applications | Flow Cytometry |
| Conjugate | CoraLite 594 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17761459
|
Proteintech
CL594-80462-20UL |
20 μL |
1651.35 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17649976
|
Proteintech
CL594-80462-100UL |
100 μL |
5307.92 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phospho-AKT1 (Ser473) antibody apecifically detetects Phospho-AKT1 (Ser473) antigen in Human, Mouse Samples. It is validated for Flow Cytometry.Specifications
| Phospho-AKT1 (Ser473) | |
| Flow Cytometry | |
| Liquid | |
| Human, Mouse | |
| NM_005163 | |
| AKT1 | |
| IgG | |
| Protein A purification |
| 2M10 | |
| CoraLite 594 | |
| Rabbit | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| 207 | |
| Peptide | |
| Primary | |
| Store at -20°C. Avoid exposure to light. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title