missing translation for 'onlineSavingsMsg'
Learn More

Phospho-AKT1 (Ser473) Rabbit anti-Human, Mouse, CoraLite 594, Clone: 2M10, Proteintech

Product Code. 17761459 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
Product Code. Quantity unitSize
17761459 20 μL 20µL
17649976 100 μL 100µL
2 options
This item is not returnable. View return policy

Product Code. 17761459

Brand: Proteintech CL5948046220UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Antibody

Phospho-AKT1 (Ser473) antibody apecifically detetects Phospho-AKT1 (Ser473) antigen in Human, Mouse Samples. It is validated for Flow Cytometry.

Specifications

Antigen Phospho-AKT1 (Ser473)
Applications Flow Cytometry
Clone 2M10
Concentration 1000 ug/mL
Conjugate CoraLite 594
Formulation PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
Gene Accession No. NM_005163
Gene Symbols AKT1
Host Species Rabbit
Immunogen Peptide
Purification Method Protein A purification
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 207
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid exposure to light.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.