missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Phillips and Hart Salt Mixture

Product Code. 11495652 Shop All MP Biomedicals Products
Click to view available options
Quantity:
2 kg
10 kg
50 kg
Unit Size:
10kg
2kg
50kg
This item is not returnable. View return policy

Product Code. 11495652

Brand: MP Biomedicals™ undefined

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Salt mixture for research animal diet.

  • Salt Mixture
  • For research animal diet
  • Powder
TRUSTED_SUSTAINABILITY

Specifications

Product Type Animal Diet
For Use With (Application) Research animal diet
Quantity 50 kg
Storage Requirements Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.