Learn More
SI Analytics™ pH Meter Lab 855
Shop All SI Analytics Products
Click to view available options
Range (pH):
2 to 16
Unit Size:
Each
Description
- Reproducible measuring results with active AutoRead function
- Simple calibration with adjustable calibration timer
- Intuitive operation with clearly arranged keyboard
- Precise measurements
- Easy to operate
Specifications
Specifications
| Range (pH) | 2 to 16 |
| Range (mV) | -2000 to 2000 |
| Temperature Range (Metric) | -10°C to +100°C |
| Display Type | LCD, Graphic, 128 x 64 pixel, Backlit |
| Model | Lab 855 |
| Channels | 1 |