Learn More
Fisherbrand™ pH-Fix Test Strips, 1.7-3.8
Product Code. 10017950 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
pH Range:
1.7 to 3.8
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Unit Size
10017950 1.7 to 3.8 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10017950
  • ONLINE EXCLUSIVE PRICE
    127.00 NOK
    92.39 NOK / Pack of 100
    Save 34.61 NOK
    27% Off
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket
Provides outstanding measurements safely over the designated pH range. Fisherbrand™ pH-Fix Test Strips, 1.7-3.8 are sufficiently long to avoid contact between the fingers and the test sample.
  • Available in a flat box so they fit into your pocket
  • Color scale on the test strip box allows exact comparison with the color developing on the test strip pad
  • Hydrophobic plastic handle protects user from sample and allows pH testing in narrow and deep vessels
  • Indicator is covalently bonded on the pads and does not bleed into the sample - can be left in the sample for an indefinite period of time without contamination
Specifications
pH Range 1.7 to 3.8
Quantity 100 Pk.
Type ph Test Strips