Includes
cell and holder
- Provides temperature-compensated pH measurements
- Made from FFKM, Ti and glass
Specifications
Specifications
| Description | For continuous pH measurement |
| For Use With (Application) | Liquid Chromatography |
| Type | pH Electrode |
| Includes | cell and holder |
| pH | 0 to 14 |