Learn More
Cytiva (Formerly GE Healthcare Life Sciences) pH Electrode
For use with ÄKTA Pilot 600
Brand: Cytiva (Formerly GE Healthcare Life Sciences) 29386679
This item is not returnable.
View return policy
Description
Specifications
Material Complies with USP Class VI | |
pH Electrode | |
Monitoring pH | |
ÄKTA Pilot 600 |