Description
- Combination electrode
- Glass shaft
- 0 to 14 pH
- Silamid™-reference system
Specifications
Specifications
| Description | pH Combination electrode |
| Material | Glass Shaft |
| pH Range | 0 to 14 |
| Membrane | Sphere |
| Model | Pl 80-225 Ph |
| Temperature (Metric) Range | 0°C to 130°C |
| Diameter (Metric) | 12 mm |