Learn More
Gilson™ Peristaltic Pump Tubing for Gilson™ MF1 and MF4 Medium Flow Pump Heads
Shop All Gilson ProductsDescription
As part of our MINIPULS™ 3 and Evolution lines of peristaltic pumps, Gilson offers a variety of tubing of different material, inner diameter sizes, and lengths.
Choice of Materials
- Silicone (Versilic™) tubing—is biologically neutral and does not inhibit cell culture while meeting European Pharmacopeia requirements, USP Class VI biological tests, and ISO 10993 biocompatibility.
- ISOVERSINIC™ tubing—shows excellent resistance to strong acids, oxidizing agents, and aromatic and chlorinated solvents.
- PharMed™ tubing—meets European Pharmacopeia requirements, USP Class VI biological tests, and ISO 10993 biocompatibility.
- PVC (Tygon™) tubing—handles virtually all inorganic chemicals found in the lab and can be used in corrosive environments.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 0.51 mm |
| Material | PVC |
| Overall Length (Metric) | 40 cm |
| For Use With | MF1 and MF4 medium flow pump head |
| For Use With (Application) | Medium flow tubing has color coded retaining stops for easy recognition of the tubing's internal diameter |
| Sterility | Sterile |
| Flow Rate | 1.42mL/min. |
| Gas Permeability | Low |
| Color | Orange/Yellow |
| Show More |