Learn More
Gilson™ Peristaltic Pump Tubing for Gilson™ MF1 and MF4 Medium Flow Pump Heads
Product Code. 10357422 Shop All Gilson Products
Click to view available options
Flow Rate:
1.42mL/min.
14mL/min.
16mL/min.
1mL/min.
2.8mL/min.
25mL/min.
29mL/min.
2mL/min.
3mL/min.
47mL/min.
55mL/min.
70mL/min.
9mL/min.
Gas Permeability:
High
Impermeable to most gases
Low
Color:
Black
Black/White
Blue
Gray
Orange/Yellow
Pink/Pink
Purple
Purple/Black
Violet/Black
Violet/White
White
Yellow/Blue
Material:
Iso-Versinic
PVC
Silicone
Unit Size:
3m
Pack of 12
Pack of 4
Product Code. 10357422
Brand: Gilson™ F1825112
Only null left
Add to basket
Add to basket
Manufactured to the utmost standard. Gilson™ Peristaltic Pump Tubing for Gilson™ MF1 and MF4 Medium Flow Pump Heads are autoclavable.
As part of our MINIPULS™ 3 and Evolution lines of peristaltic pumps, Gilson offers a variety of tubing of different material, inner diameter sizes, and lengths.
Choice of Materials
  • Silicone (Versilic™) tubing—is biologically neutral and does not inhibit cell culture while meeting European Pharmacopeia requirements, USP Class VI biological tests, and ISO 10993 biocompatibility.
  • ISOVERSINIC™ tubing—shows excellent resistance to strong acids, oxidizing agents, and aromatic and chlorinated solvents.
  • PharMed™ tubing—meets European Pharmacopeia requirements, USP Class VI biological tests, and ISO 10993 biocompatibility.
  • PVC (Tygon™) tubing—handles virtually all inorganic chemicals found in the lab and can be used in corrosive environments.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Diameter (Metric) Inner 1 mm
Material Silicone
Overall Length (Metric) 40 cm
For Use With MF1 and MF4 medium flow pump head
For Use With (Application) Medium flow tubing has color coded retaining stops for easy recognition of the tubing's internal diameter
Sterility Sterile
Flow Rate 1mL/min.
Gas Permeability High
Color White
Product Type Peristaltic Pump Tubing