missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Periostin Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | Periostin |
|---|---|
| Concentration | 200 μg/mL |
| Applications | Immunohistochemistry, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16845835
|
Proteintech
27794-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16835835
|
Proteintech
27794-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Periostin antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Periostin. This antibody recognizes human antigen. The Periostin antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| Periostin | |
| Immunohistochemistry, ELISA | |
| Unconjugated | |
| Human | |
| IgG |
| 200 μg/mL | |
| Polyclonal | |
| Rabbit | |
| 10631 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title