Learn More
Periostin Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27794-1-AP-150UL
Description
The Periostin antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Periostin. This antibody recognizes human antigen. The Periostin antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| Periostin | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 10631 | |
| IgG |
| Immunohistochemistry, ELISA | |
| 200 μg/mL | |
| Rabbit | |
| Primary | |
| Human |