Learn More
Periostin Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27794-1-AP-150UL
 View more versions of this product
Product Code. 16835835
  • 4836.10 NOK / 150µL
Description
The Periostin antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Periostin. This antibody recognizes human antigen. The Periostin antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
Periostin
Polyclonal
Unconjugated
150 μL
10631
IgG
Immunohistochemistry, ELISA
200 μg/mL