Learn More
Pepsin/Hydrochloric Acid Electrode Cleaning Solution, SI Analytics™
Product Code. 10213511 Shop All SI Analytics Products
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1 L
250mL
Product Code. 10213511
Brand: SI Analytics 285138538
SI Analytics™ Pepsin/Hydrochloric Acid Electrode Cleaning Solution is a corrosive solution that is suitable for cleaning electrodes.
  • Water, pepsin A, hydrochloric acid
  • Classification according to Regulation (EC) No 1272/2008: HS05 corrosion
  • The product does not contain any relevant quantities of materials with critical values that have to be monitored at the workplace
  • Avoid contact with the eyes and skin
  • Do not inhale gases / fumes / aerosols
  • Take off contaminated clothing and wash it before reuse
  • Formation of hydrogen possible with metals and alloys (risk of explosion)
  • Generally not hazardous for water
TRUSTED_SUSTAINABILITY
Specifications
Boiling Point 100°C
Color Colorless
Filling Solution Type Cleaning Solution
Odor Odorless
pH Range 1
Quantity 1000 mL
Solubility Fully miscible in water
Vapor Pressure 23hPa
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • EUH208-Contains pepsin A. May produce an allergic reaction.
Precautionary Statement
  • P234-Keep only in original container.
Supplemental information
  • MIXTURE LIST-Contains : pepsin A