Learn More
Memmert™ Entry Port
Product Code. 15312732 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
IPP30
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
15312732 IPP30 Each
1 options
Product Code. 15312732
Brand: Memmert™ IPP30F0
Only null left
Add to basket
Add to basket
Material: Silicone; For Use With: IPP30
  • 23 mm clear diameter
  • For introducing connections at the side
  • Can be closed by flap
  • Position left centre/centre or centre top
Specifications
For Use With (Equipment) IPP30
Material Silicone
Type Entry Port
Diameter (Metric) Inside 23 mm