Learn More
Eppendorf™ PCR-Cooler
Product Code. 10192281 Shop All Eppendorf Products
Change view
Click to view available options
Color:
Blue
Blue, Pink
Pink
Unit Size:
1 set
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10192281 Blue Each
10275091 Pink Each
10013051 Blue, Pink 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10192281 Supplier Eppendorf™ Supplier No. 3881000031
Sample Preparation and Storage Essentials
Use promo code 27996
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
Innovative solution for protecting, transporting and storing sensitive samples as well as for safely stopping reactions.
  • Maintains a sample temperature of 0°C for over an hour and then changes color when the temperature has exceeded 7°C
  • Superior to an ice bath, it is easy to handle, space saving and free of contamination, which makes it the perfect preparation for PCR reactions
  • Temperature of single tubes is indicated by the color of the individual bores
  • Samples are prevented from freezing by the specially developed cone contour
  • Cooling aggregate in the universal 96 well format for 0.2mL and 0.5mL PCR tubes, for tube strips, plates and individual tubes
TRUSTED_SUSTAINABILITY
Spécification
For Use With (Application) PCR Reaction Setup
Type PCR Cooler
Color Blue
Capacity (Metric) 0.2 mL
Holds 96
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.