Learn More
Ratiolab™ PC Cryo Boxes
Made of high quality colored polycarbonate (PC) for use with liquid nitrogen.
762.00 NOK - 947.00 NOK
Specifications
| Material | Polycarbonate |
|---|---|
| Shape | Square |
| Type | Cryo Box |
| For Use With (Application) | Storage and Transport with Liquid Nitrogen |
| For Use With (Equipment) | Cryo tubes 1.2-2 mL |
| Product Code | Brand | Color | Dimensions (L x W x H) | Tube Array | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Color | Dimensions (L x W x H) | Tube Array | Price | Quantity & Availability | |||
|
15941651
|
Ratiolab™
5150002 |
Green | 132 x 132 x 52 mm | 9 x 9 grid |
762.00 NOK
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
|
15942551
|
Ratiolab™
5150031 |
Blue | 76 x 76 x 52 mm | 5 x 5 grid |
947.00 NOK
Pack of 8 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Autoclavable and temperature resistant from -190° to +121°C
- 81 or 100 places for 9 x 9 and 10 x 10 grids
- 1/4-format with 25 places, 5 x 5 grid
- Numerically coded
- Stackable for safe transport
- Drain bores at the bottom for dew liquids
Specifications
| Polycarbonate | |
| Cryo Box | |
| Cryo tubes 1.2-2 mL | |
| -190°C | |
| 52 mm |
| Square | |
| Storage and Transport with Liquid Nitrogen | |
| 121°C | |
| -190°C to +121°C |