Learn More
Ratiolab™ PC Cryo Boxes
Product Code. 15942551
Change view
Click to view available options
Color:
Blue
Green
Dimensions (L x W x H):
76 x 76 x 52 mm
132 x 132 x 52 mm
Tube Array:
5 x 5 grid
9 x 9 grid
Unit Size:
Pack of 5
Pack of 8
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Dimensions (L x W x H) Tube Array unitSize
15942551 Blue 76 x 76 x 52 mm 5 x 5 grid Pack of 8
15941651 Green 132 x 132 x 52 mm 9 x 9 grid Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15942551 Supplier Ratiolab™ Supplier No. 5150031
Made of high quality colored polycarbonate (PC) for use with liquid nitrogen.
  • Autoclavable and temperature resistant from -190° to +121°C
  • 81 or 100 places for 9 x 9 and 10 x 10 grids
  • 1/4-format with 25 places, 5 x 5 grid
  • Numerically coded
  • Stackable for safe transport
  • Drain bores at the bottom for dew liquids
Specifications
Color Blue
Material Polycarbonate
Shape Square
Type Cryo Box
For Use With (Application) Storage and Transport with Liquid Nitrogen
For Use With (Equipment) Cryo tubes 1.2-2 mL
Holds 25 (1.2 mL and 2 mL Cryogenic Tubes)
Max. Temperature (Metric) 121°C
Min. Temperature (Metric) -190°C
Quantity 8/pk
Tube Array 5 x 5 grid
Withstand Temperature (Metric) -190°C to +121°C
Dimensions (L x W x H) 76 x 76 x 52 mm
Length (Metric) 76 mm
Width (Metric) 76 mm
Height (Metric) 52 mm