Learn More
Amarell™ Partial Immersion Eco-friendly Thermometer
Specifications
Specifications
| Bottle Fill | Green Liquid |
| For Use With (Application) | General use |
| Immersion Type | Partial Immersion |
| Length (Metric) | 300 mm |
| Product Type | Partial Immersion Thermometer |
| Max. Temperature (Metric) | 110°C |
| Min. Temperature (Metric) | -10°C |
| Temperature Range (Metric) | -10°C to +110°C |
| Subdivision | 0.5°C |
| Immersion Length | 76 mm |