Learn More
Pancreatic Polypeptide Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | Pancreatic Polypeptide |
|---|---|
| Concentration | 133 μg/mL |
| Applications | Immunohistochemistry, Immunofluorescence, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16894694
|
Proteintech
15493-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16884694
|
Proteintech
15493-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Pancreatic Polypeptide antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Pancreatic Polypeptide. This antibody recognizes human, mouse, rat antigen. The Pancreatic Polypeptide antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
| Pancreatic Polypeptide | |
| Immunohistochemistry, Immunofluorescence, ELISA | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG |
| 133 μg/mL | |
| Polyclonal | |
| Rabbit | |
| 5539 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.