missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pancreatic Polypeptide Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Shop All Proteintech ProductsDescription
The Pancreatic Polypeptide antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Pancreatic Polypeptide. This antibody recognizes human, mouse, rat antigen. The Pancreatic Polypeptide antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
Specifications
| Antigen | Pancreatic Polypeptide |
| Applications | Immunohistochemistry, Immunofluorescence, ELISA |
| Classification | Polyclonal |
| Concentration | 133 μg/mL |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Quantity | 150 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 5539 |
| Target Species | Human, Rat, Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?