Learn More
Pancreatic Polypeptide Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15493-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The Pancreatic Polypeptide antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Pancreatic Polypeptide. This antibody recognizes human, mouse, rat antigen. The Pancreatic Polypeptide antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
Pancreatic Polypeptide | |
Polyclonal | |
Unconjugated | |
20 μL | |
5539 | |
IgG |
Immunohistochemistry, Immunofluorescence, ELISA | |
133 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |