Learn More
Pancreatic Polypeptide Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15493-1-AP-20UL
 View more versions of this product
Product Code. 16894694
  • 1710.33 NOK / 20µL
Description
The Pancreatic Polypeptide antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Pancreatic Polypeptide. This antibody recognizes human, mouse, rat antigen. The Pancreatic Polypeptide antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
Specifications
Pancreatic Polypeptide
Polyclonal
Unconjugated
20 μL
5539
IgG
Immunohistochemistry, Immunofluorescence, ELISA
133 μg/mL
Rabbit
Primary
Human, Rat, Mouse