missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PADI3 Monoclonal Antibody (C10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543631
This item is not returnable.
View return policy
Description
PADI3 Monoclonal antibody specifically detects PADI3 in Human, Rat samples. It is validated for Western Blot.
Specifications
| PADI3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P70708, Q9ULW8 | |
| PADI3 | |
| Met1-Pro300 (Accession Q9ULW8), with N-terminal His Tag. | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2b κ |
| Western Blot | |
| C10 | |
| Unconjugated | |
| PADI3 | |
| PAD3; PADI3; PDI3; peptidyl arginine deiminase 3; peptidyl arginine deiminase, type III; peptidylarginine deiminase III; Protein-arginine deiminase type III; protein-arginine deiminase type-3 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 29520, 51702 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction