missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PADI3 Monoclonal Antibody (C10), Invitrogen™

Mouse Monoclonal Antibody
2060.00 NOK - 4515.00 NOK
Specifications
| Antigen | PADI3 |
|---|---|
| Clone | C10 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17674086
|
Invitrogen™
MA543631 |
20 μL |
2060.00 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17664086
![]() |
Invitrogen™
MA543630 |
100 μL |
4515.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PADI3 Monoclonal antibody specifically detects PADI3 in Human, Rat samples. It is validated for Western Blot.Specifications
| PADI3 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| PAD3; PADI3; PDI3; peptidyl arginine deiminase 3; peptidyl arginine deiminase, type III; peptidylarginine deiminase III; Protein-arginine deiminase type III; protein-arginine deiminase type-3 | |
| PADI3 | |
| IgG2b κ | |
| Protein A/G | |
| Antibody |
| C10 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| P70708, Q9ULW8 | |
| 29520, 51702 | |
| Met1-Pro300 (Accession Q9ULW8), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| PADI3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title