Description
Use Thermo Scientific™ μPAC™ ESI-MS Accessories to facilitate reproducible, low dispersion, and grounded connections between μPAC Neo Plus HPLC columns and low-flow mass spectrometer ion sources. μPAC Neo Plus voltage spacers are available for both EASY-Spray and Nanospray Flex ion sources. All configurations require a grounding cable.
The μPAC Flex iON Connect ESI-MS Interface consists of two separate units:
- A docking unit that can be slid onto the Nanospray Flex Ion Source's metal rod and receives both a grounding and a high voltage cable
- A spray unit compatible with the outlet fittings of μPAC columns on one side and compatible with any 360 μm OD emitter on the other side.
The μPAC EASY-Spray emitter is designed for easy connection to an EASY-Spray MS Source. It contains a pre-assembled EASY-Spray emitter with a transfer line and grounding union.
Specifications
Specifications
| Description | μPAC™ Flex iON Connect ESI-MS Interface |
| For Use With (Equipment) | Nanospray Flex™ Ion Source |
| Type | Interface |
| Length (Metric) | N/A |
| Diameter (Metric) Inner | N/A |