missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p63 Mouse anti-Human, Unconjugated, Clone: 2B6B2, Proteintech
Mouse Monoclonal Antibody
1660.20 NOK - 4731.57 NOK
Specifications
| Antigen | p63 |
|---|---|
| Clone | 2B6B2 |
| Concentration | 633 μg/mL |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Monoclonal |
Description
The p63 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human p63. This antibody recognizes human antigen. The p63 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| p63 | |
| 633 μg/mL | |
| Monoclonal | |
| Mouse | |
| 8626 | |
| Primary |
| 2B6B2 | |
| Western Blot, Immunohistochemistry, ELISA | |
| Unconjugated | |
| Human | |
| IgG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title