Learn More
p63 Mouse anti-Human, Unconjugated, Clone: 2B6B2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 60332-1-IG-150UL
 View more versions of this product
Product Code. 16837435
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
The p63 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human p63. This antibody recognizes human antigen. The p63 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
p63
Monoclonal
633 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
2B6B2
Unconjugated
150 μL
8626
IgG1